Learn More
Helix Medical Inc TUBING SIL .125 X .250 50FT
Catalog No. NC9459570
Encompass_Preferred
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.
This item is not returnable. View return policy
Catalog No. NC9459570 Supplier Helix Medical Inc Supplier No. 6001121
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.