Learn More
Helix Medical Inc TUBING SIL .125 X .251 25FT
Catalog No. NC0090775
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.
Catalog No. NC0090775 Supplier Helix Medical Inc Supplier No. 6082579
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.