Learn More
Helix Medical Inc TUBING SIL .125 X .313 25FT
Catalog No. NC9036407
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.
Catalog No. NC9036407 Supplier Helix Medical Inc Supplier No. 6082582
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.