Learn More
Helix Medical Inc TUBING SIL .188 X .375 50FT
Supplier:  Helix Medical Inc 6041153
Encompass_Preferred
Biomedical/pharmaceutical grade standard tubing, made with Wacker Silicones HM-6540 platinum-cured material. The manufacturing process minimizes flow degradation over extended periods. Meets USP Class VI biocompatibility requirements.
Catalog No. NC0476808