Learn More
Helix Medical Inc TUBING SIL .250 X .375 50FT
Catalog No. NC9251375
Encompass_Preferred
HelixMark Silicone Tubing, 50 ft. Coils, Inside dia.: .250 in., Outside dia.: .375 ft.
Catalog No. NC9251375 Supplier Helix Medical Inc Supplier No. 6001124
HelixMark Silicone Tubing, 50 ft. Coils, Inside dia.: .250 in., Outside dia.: .375 ft.