Learn More
Helix Medical Inc TUBING SIL .250 X .376 25FT
Catalog No. NC0090776
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.
Catalog No. NC0090776 Supplier Helix Medical Inc Supplier No. 60825130
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing.