Learn More
Helix Medical Inc Standard Silicone Tubing .250inch (ID) x .438inch (OD) 50ft
Catalog No. NC0065976
Encompass_Preferred
Please note this is a customer item and non returnable. Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.
Catalog No. NC0065976 Supplier Helix Medical Inc Supplier No. 6001125
Please note this is a customer item and non returnable. Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.