Learn More
Helix Medical Inc TUBING SIL .250 X .450 25FT
Catalog No. NC0090778
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing. Fits Sz 24TW.
Catalog No. NC0090778 Supplier Helix Medical Inc Supplier No. 60825136
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing. Fits Sz 24TW.