Learn More
Helix Medical Inc TUBING SIL .250 X .500 50FT
Catalog No. NC0147745
Encompass_Preferred
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.
Catalog No. NC0147745 Supplier Helix Medical Inc Supplier No. 6001126
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.