Learn More
Helix Medical Inc Dow Corning Silicone Tubing, 0.375in ID x 0.500in OD, 50ft
Catalog No. NC1544827
Encompass_Preferred
HelixMark standard silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fl uids under standard requirements.
This item is not returnable. View return policy
Catalog No. NC1544827 Supplier Helix Medical Inc Supplier No. 6001128
HelixMark standard silicone tubing meets the needs of a broad range of applications and is universally applicable for the transfer of fl uids under standard requirements.