Learn More
Helix Medical Inc TUBING SIL .375 X .625 50FT
Catalog No. NC9082662
Encompass_Preferred
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.
Catalog No. NC9082662 Supplier Helix Medical Inc Supplier No. 6001130
Platinum cured, non-implantable, non-adherent tubing for bio/pharma industry standards. Ability to operate at elevated temperatures. Will not oxidize/deteriorate. Can be sterilized with autoclave, ethylene oxide, or gamma.