Learn More
Helix Medical Inc TUBING SIL .375 X .645 25FT
Catalog No. NC0090780
Encompass_Preferred
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing. Fits Sz 73TW.
Catalog No. NC0090780 Supplier Helix Medical Inc Supplier No. 60825197
Platinum-cured 60 duro peristaltic tubing that provides superior strength, performance and longer life. Meets strength and durability requirements associated with peroxide-cured tubing. Fits Sz 73TW.