Learn More
Helix Medical Inc TUBING SIL .500 X .750 50FT
Catalog No. NC0237182
Encompass_Preferred
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.
Catalog No. NC0237182 Supplier Helix Medical Inc Supplier No. 6001132
Platinum-cured peristaltic pump tubing, manufactured with Wacker Silicones HM-6540 material, has the strength and durability normally associated with peroxide-cured tubing.