Learn More
Chemglass Life Sciences Tubing, Silicone, Peroxide-Cured Silcon, Size 16, 1/8" ID x 1/4" OD x 1/16" Wall Thickness x 50' Long
SDP
Catalog No. 50143760
Encompass_Preferred
Peroxide-cured Silcon Silicone Tubing is designed for use in applications where flexibility, resiliency and durability are required. Its inner smooth surface reduces the risk of particulate entrapment and microscopic buildup during fluid transfer.Tubing is FDA food grade compliant, entirely non-toxic, non-hemolytic and non-pyrogenic. It also meets 3-A Sanitary Standards, FDA 21 CFR Part 177.2600 Criteria and NSF 51 Standard. Withstands temperature extremes from -100°F to 500°F (350°F maximum for NSF applications).May be low pressure steam sterilized in-line or autoclaved at up to 121°C in a normal autoclaving cycle. May be sterilized repeatedly but if tubing becomes relaxed or gummy it should be replaced. Sold in standard lengths of 50ft.
Catalog No. 50-143-760 Supplier Chemglass Life Sciences Supplier No. CLS1436T20
Peroxide-cured Silcon Silicone Tubing is designed for use in applications where flexibility, resiliency and durability are required. Its inner smooth surface reduces the risk of particulate entrapment and microscopic buildup during fluid transfer.Tubing is FDA food grade compliant, entirely non-toxic, non-hemolytic and non-pyrogenic. It also meets 3-A Sanitary Standards, FDA 21 CFR Part 177.2600 Criteria and NSF 51 Standard. Withstands temperature extremes from -100°F to 500°F (350°F maximum for NSF applications).May be low pressure steam sterilized in-line or autoclaved at up to 121°C in a normal autoclaving cycle. May be sterilized repeatedly but if tubing becomes relaxed or gummy it should be replaced. Sold in standard lengths of 50ft.