Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Peroxide-Cured Silicone, 1/2"" ID x 3/4"" OD; 25 ft
Catalog No. NC1667876
Encompass
Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications. Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -60 to 460¿F (-51 to 238¿C) Sterilize: Autoclave or gamma irradiation.
Catalog No. NC1667876 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0641155
Applications: Pharmaceutical, biotechnology, food and beverage, aerospace, and general lab applications. Characteristics: Peroxide-cured, excellent heat resistance, low compression set, odorless, nontoxic, translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and meets USP Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -60 to 460¿F (-51 to 238¿C) Sterilize: Autoclave or gamma irradiation.