Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Platinum-Cured Silicone, 1/16"" ID x 1/8"" OD; 25 ft
Catalog No. NC9656699
Encompass
No need to worry about the heat - use for applications up to 460F. Ideal for general-purpose, single-use, and extended fluid path applications. Global market leader in range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime.
Catalog No. NC9656699 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9580202
No need to worry about the heat - use for applications up to 460F. Ideal for general-purpose, single-use, and extended fluid path applications. Global market leader in range of formulations for most extensive chemical compatibility. Single-use custom lengths and configurations available to minimize downtime.