Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon 3370 I.B. Reinforced Platinum-Cured Silicone, 3/8"" ID x 5/8"" OD; 50 ft
Catalog No. 501961046
Encompass
Formulation: Tygon 3370 I. B., silicone, platinum cured. Applications: Demanding sanitary high-pressure pharmaceutical, biotechnology, food and beverage, and cosmetic applications. Characteristics: Excellent heat resistance, low compression set, odorless, and nontoxic. Translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and USP Class V extractables; exceeds Class VI implant; exceeds 3A Sanitary Standards; produced to Bulk Pharmaceutical cGMPs (FDA 21 CFR 210, 211). Tubing manufactured according to GMP Temperature range: ¿60 to 320¿F (¿51 to 160¿C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation.
Catalog No. 50-196-1046 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9570302
Formulation: Tygon 3370 I. B., silicone, platinum cured. Applications: Demanding sanitary high-pressure pharmaceutical, biotechnology, food and beverage, and cosmetic applications. Characteristics: Excellent heat resistance, low compression set, odorless, and nontoxic. Translucent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600) and USP Class V extractables; exceeds Class VI implant; exceeds 3A Sanitary Standards; produced to Bulk Pharmaceutical cGMPs (FDA 21 CFR 210, 211). Tubing manufactured according to GMP Temperature range: ¿60 to 320¿F (¿51 to 160¿C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation.