Learn More
Chemglass Life Sciences Tubing, Silicone, Silcon Med-X Platinum-Cured, Size 120, 5/16" OD x 1/8" ID x 3/32" Wall Thickness x 50' Long
SDP
Catalog No. 5000083196
Encompass_Preferred
Tubing, Silicone, Platinum-Cured Versilic, Size 120, 5/16" OD x 1/8" ID x 3/32" Wall Thickness x 50' Long
Catalog No. 50-000-83196 Supplier Chemglass Life Sciences Supplier No. CLS1436P03
Tubing, Silicone, Platinum-Cured Versilic, Size 120, 5/16" OD x 1/8" ID x 3/32" Wall Thickness x 50' Long