Learn More
Chemglass Life Sciences Tubing, Silicone, Silcon Med-X Platinum-Cured, Size 14, 3/16" OD x 1/16" ID x 1/16" Wall Thickness x 50' Long
SDP
Catalog No. 5000083134
Encompass_Preferred
Raw materials meet USP Class VI requirements Manufactured under good manufacturing procedures in a controlled environment Platinum cured for the highest degree of purity (least extractables) Reusable - will withstand repeated sterilization Translucent natural color for visual contact with the flow Applications include pharmaceutical, biomedical, health and beauty, food, and beverage handling Free of BPA, latex and phthalates RoHS compliant tubing is made under good manufacturing procedures and conforms to the biocompatibility requirements of USP Class VI.surface properties resist sticking and encrustation and will not support bacteria growth.The tubing is soft, pliable, and contains no plasticizers which could leach out, causing flow contamination or tube hardening.Care is recommended in the selection and application of fittings and clamps as sharp barbed fittings or unlined metal clamps could tear into the wall.
Catalog No. 50-000-83134 Supplier Chemglass Life Sciences Supplier No. CLS1436P01
Raw materials meet USP Class VI requirements Manufactured under good manufacturing procedures in a controlled environment Platinum cured for the highest degree of purity (least extractables) Reusable - will withstand repeated sterilization Translucent natural color for visual contact with the flow Applications include pharmaceutical, biomedical, health and beauty, food, and beverage handling Free of BPA, latex and phthalates RoHS compliant tubing is made under good manufacturing procedures and conforms to the biocompatibility requirements of USP Class VI.surface properties resist sticking and encrustation and will not support bacteria growth.The tubing is soft, pliable, and contains no plasticizers which could leach out, causing flow contamination or tube hardening.Care is recommended in the selection and application of fittings and clamps as sharp barbed fittings or unlined metal clamps could tear into the wall.