Learn More
Chemglass Life Sciences Tubing, Silicone, Silcon Med-X Platinum-Cured, Size 26, 1/2" OD x 1/4" ID x 1/8" Wall Thickness x 50' Long
SDP
Catalog No. 5000083098
Encompass_Preferred
Tubing, Silicone, Platinum-Cured Versilic, Size 26, 1/2" OD x 1/4" ID x 1/8" Wall Thickness x 50' Long
Catalog No. 50-000-83098 Supplier Chemglass Life Sciences Supplier No. CLS1436P08
Tubing, Silicone, Platinum-Cured Versilic, Size 26, 1/2" OD x 1/4" ID x 1/8" Wall Thickness x 50' Long