Learn More
IDEX Health & Science Upchurch Tubing Sleeve FEP Green 1/16" OD x .0155" ID x 2" Green - Single
Catalog No. NC1209552
Encompass_Preferred
This item has a minimum order quantity of 25 per supplier requirements. Tubing Sleeve FEP Green 1/16" OD x .0155" ID x 2" Green - Single
Catalog No. NC1209552 Supplier IDEX Health & Science Upchurch Supplier No. F262
This item has a minimum order quantity of 25 per supplier requirements. Tubing Sleeve FEP Green 1/16" OD x .0155" ID x 2" Green - Single