Learn More
Microsolv Technology Corporation Tubing, Stainless Steel, Pre-Cut, 0.005" ID, 1/16" OD, HPLC Grade, Red Band, 30cm Long. 1 EA.
Catalog No. 502271101
Encompass_Preferred
Properly cutting tubing is one of the most difficult tasks in a chromatography laboratory. These Pre-Cut Stainless Steel Tubes are the perfect solution.Welded and Annealed, the ends are cut using Electronic Discharge ( EDC ) Technique to achieve square and burr free zero dead volume connections. They are tested according to EN 10204/3.1B. The Red Band is a color code for.005" ID. Internal and Outer Dimension Tolerances: +/- 0.05mm.
This item is not returnable. View return policy
Catalog No. 50-227-1101 Supplier Microsolv Technology Corporation Supplier No. 4940530
Properly cutting tubing is one of the most difficult tasks in a chromatography laboratory. These Pre-Cut Stainless Steel Tubes are the perfect solution.Welded and Annealed, the ends are cut using Electronic Discharge ( EDC ) Technique to achieve square and burr free zero dead volume connections. They are tested according to EN 10204/3.1B. The Red Band is a color code for.005" ID. Internal and Outer Dimension Tolerances: +/- 0.05mm.