Learn More
Med Vet International Dynarex Oxygen Supply Tubing, 14' (4.3 M) Crush Resistant, Universal Connector, 50/Case
Catalog No. 502832417
Encompass_Preferred
Dynarex Resp-O2 Oxygen Supply Tubing is designed to extend a patient’s reach when accessing a concentrated oxygen source for respiratory therapy. The tubing features a light green color, designed to help improve tube visibility. It comes with a standard connector on one end and either a standard or universal connector on the other. Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP
Catalog No. 50-283-2417 Supplier Med Vet International Supplier No. 33203
Dynarex Resp-O2 Oxygen Supply Tubing is designed to extend a patient’s reach when accessing a concentrated oxygen source for respiratory therapy. The tubing features a light green color, designed to help improve tube visibility. It comes with a standard connector on one end and either a standard or universal connector on the other. Crush-resistant, star lumen oxygen supply tubing Individually wrapped Not made with natural rubber latex or DEHP