Learn More
Waters Corp Waters Tubing Assembly, T1, QSM to V2 Port 5
Catalog No. 50466552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is designed for connecting the QSM to V2 port 5 in a T1 configuration. It is categorized as a spare part kit, ensuring compatibility and reliable performance for maintenance and replacement needs.
  • Designed for T1, QSM to V2 port 5 connections.
  • Categorized as a spare parts kit.
Catalog No. 50-466-552 Supplier Waters Corp Supplier No. 430002517
This tubing assembly is designed for connecting the QSM to V2 port 5 in a T1 configuration. It is categorized as a spare part kit, ensuring compatibility and reliable performance for maintenance and replacement needs.
  • Designed for T1, QSM to V2 port 5 connections.
  • Categorized as a spare parts kit.