Learn More
Cole-Parmer Reinforced TPE Tubing, Clear, 1/4" ID x 1/2" OD; 25 Ft
Supplier:  Cole-Parmer 5011631
Encompass_Preferred
The Cole-Parmer Reinforced Thermoplastic Vulcanizate (TPV) tubing is soft and flexible tubing that is durable enough to provide a long service life. This tubing is designed for use with peristaltic pumps. This TPE tubing does not contain PVC, DEHP, phthalates, or plasticizers, making them ideal for various pharmaceutical, medical, food, and beverage applications. The reinforced tubing design makes it ideal for demanding, high pressure use.
Catalog No. 50-255-7901