Learn More
Bellco Glass, Inc. Tubing, TPE, EZBio, 1/4" ID x 3/8" OD, with Text, 50ft/Roll
SDP
Catalog No. 502550122
Encompass_Preferred
EZBio Thermoplastic elastomers tubing allows easy replacement or extension. EZBio tubing allows users to replace or extend the tubing connecting their Vactrap vacuum trap system (sold separately).to their vacuum. 50-foot roll allows you to make custom extensions to improve Vactrap versatility and mobility in the lab. Includes a replacement inline vent filter. See replacement vent filters. Say goodbye to glass. Say hello to Vactrap . SPECIFICATIONSSKU/Product Code: 25T-3418-FLSManufacturer: Material: Thermoplastic elastomers tubing Size/Dimensions: 1/4" ID x 3/8" OD, 50' L
Catalog No. 50-255-0122 Supplier Bellco Glass, Inc. Supplier No. 452800640
EZBio Thermoplastic elastomers tubing allows easy replacement or extension. EZBio tubing allows users to replace or extend the tubing connecting their Vactrap vacuum trap system (sold separately).to their vacuum. 50-foot roll allows you to make custom extensions to improve Vactrap versatility and mobility in the lab. Includes a replacement inline vent filter. See replacement vent filters. Say goodbye to glass. Say hello to Vactrap . SPECIFICATIONSSKU/Product Code: 25T-3418-FLSManufacturer: Material: Thermoplastic elastomers tubing Size/Dimensions: 1/4" ID x 3/8" OD, 50' L