Learn More
Hach Company Tubing, Transmission, Large, 0.060 in (1.5 mm) I.D., 50 ft (15.24 m)
Catalog No. NC2088478 Shop All Hach Company Products
Encompass
Flexible transmission tubing designed for analyzer sampling and fluid transfer applications. Supplied as a 50 ft (15.24 m) roll with a 0.060 in (1.5 mm) inner diameter. The tubing offers chemical resistance and flexibility for use in transmission lines.
  • 0.060 in (1.5 mm) inner diameter.
  • 50 ft (15.24 m) continuous roll.
  • Flexible, chemical-resistant construction.
  • Designed for transmission lines and analyzer sampling.
  • Compatible with standard tubing fittings.
This item is not returnable. View return policy
Catalog No. NC2088478 Supplier Hach Company Supplier No. 50029
Flexible transmission tubing designed for analyzer sampling and fluid transfer applications. Supplied as a 50 ft (15.24 m) roll with a 0.060 in (1.5 mm) inner diameter. The tubing offers chemical resistance and flexibility for use in transmission lines.
  • 0.060 in (1.5 mm) inner diameter.
  • 50 ft (15.24 m) continuous roll.
  • Flexible, chemical-resistant construction.
  • Designed for transmission lines and analyzer sampling.
  • Compatible with standard tubing fittings.