Learn More
AVANTOR MASTERFLEX (VWR LLC) TUBING TYGN LMT55 3.2 15M 50FT
Catalog No. 502742819
Encompass
Masterflex Ismatec Pump Tubing, Tygon LMT-55, 3.2 mm ID x 6.4 mm OD; 50 ft, Avantor
Catalog No. 50-274-2819 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0650705
Masterflex Ismatec Pump Tubing, Tygon LMT-55, 3.2 mm ID x 6.4 mm OD; 50 ft, Avantor