Learn More
Grainger Tubing: Tygon B-44-5X IB, Tygon, 1/2 in Inside Dia, 3/4 in Outside Dia, 50 ft Lg, Clear
Catalog No. NC3251697
Encompass_Preferred
Tubing: Tygon B-44-5X IB, Tygon, 1/2 in Inside Dia, 3/4 in Outside Dia, 50 ft Lg, Clear
Catalog No. NC3251697 Supplier Grainger Supplier No. 786VK9
Tubing: Tygon B-44-5X IB, Tygon, 1/2 in Inside Dia, 3/4 in Outside Dia, 50 ft Lg, Clear