Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Tygon E-3603, Non-DEHP, 1"" ID x 1-3/8"" OD; 50 ft
Catalog No. 501961030
Encompass
Formulation: Tygon E-3603 Applications: General laboratory use. Characteristics: Long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic. Transparent. Certifications: FDA compliant. Made according to GMP. Temperature range: ¿58 to 165¿F (¿50 to 74¿C) Sterilize: Ethylene oxide or autoclave for 30 min at 250¿F (121¿C), 15 psi. After autoclaving, dry at no more than 150¿F (66¿C) for 2 to 2-1/2 hours.
Catalog No. 50-196-1030 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640707
Formulation: Tygon E-3603 Applications: General laboratory use. Characteristics: Long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic. Transparent. Certifications: FDA compliant. Made according to GMP. Temperature range: ¿58 to 165¿F (¿50 to 74¿C) Sterilize: Ethylene oxide or autoclave for 30 min at 250¿F (121¿C), 15 psi. After autoclaving, dry at no more than 150¿F (66¿C) for 2 to 2-1/2 hours.