Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon Silicone Tubing, 1/4""ID x 3/8""OD, 50 ft/pack
Catalog No. NC1616196
Encompass
Formulation: TYGON 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications. Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VI requirements, NSF criteria and EP 3.2.9 requirements. Tubing is manufactured according to GMP. Temperature range: ¿112 to 392¿F (¿80 to 200¿C) Sterilize: With ethylene oxide, gamma irradiation, or autoclave for 30 min, 15 psi.
Catalog No. NC1616196 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9570213
Formulation: TYGON 3350 silicone, platinum cured. Applications: Pharmaceutical, cosmetic, industrial, and auto-analysis applications. Characteristics: Ultra-smooth inner surface reduces particle entrapment and minimizes bacterial growth. Good for mild to medium concentration bases, salts, and alcohols; is odorless, tasteless, and non-toxic. Transparent. Certification: Elastomer is FDA-compliant (21 CFR 177.2600); meets USP Class VI requirements, NSF criteria and EP 3.2.9 requirements. Tubing is manufactured according to GMP. Temperature range: ¿112 to 392¿F (¿80 to 200¿C) Sterilize: With ethylene oxide, gamma irradiation, or autoclave for 30 min, 15 psi.