Learn More
Bellco Glass, Inc. Tubing, Tygon, Silicone 3350, 1/8ID x 1/4OD, 1/EA
SDP
Catalog No. 502550120
Encompass_Preferred
Tubing is autoclavable and made from Silicone. It complies with with USP XXII Class VI criteria, FDA 21 CFR Part 177.2600 and NSF 51 Standard. Applications include, but not limited to biotechnological, pharmaceutical, and peristaltic application.SpecificationsMaterial: Silicone Color: TranslucentID: 1/8"OD: 1/4"Length: 50 ft.Wall thickness: 1/16"Certifications/Compliance: USP, FDA, NSF
Catalog No. 50-255-0120 Supplier Bellco Glass, Inc. Supplier No. 452800642
Tubing is autoclavable and made from Silicone. It complies with with USP XXII Class VI criteria, FDA 21 CFR Part 177.2600 and NSF 51 Standard. Applications include, but not limited to biotechnological, pharmaceutical, and peristaltic application.SpecificationsMaterial: Silicone Color: TranslucentID: 1/8"OD: 1/4"Length: 50 ft.Wall thickness: 1/16"Certifications/Compliance: USP, FDA, NSF