Learn More
Accuflow Systems Saint-Gobain Tygon Tubing, Thermoplastic, Vacuum, Phthalate-Free
Catalog No. 502365905
Encompass_Preferred
Tygon E-3603 is a flexible, crystal-clear thermoplastic tubing engineered for laboratory and medical vacuum and fluid transfer applications. It offers broad chemical compatibility, a glassy-smooth inner bore to minimize buildup and carryover, and vacuum performance suitable for many analytical and diagnostic systems. Available sizes and vacuum ratings vary by specific dimension; consult the manufacturer's specifications for exact values.
  • Crystal-clear for visual inspection and bubble detection.
  • Suitable for vacuum applications up to 29.9 inHg (760 mmHg) for many sizes.
  • Thermoplastic material, non-oxidizing and non-contaminating.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Available in a wide range of inner and outer diameters; refer to specifications for exact dimensions.
  • Phthalate- and BPA-free formulation.
Catalog No. 50-236-5905 Supplier Accuflow Systems Supplier No. ACF00002
Tygon E-3603 is a flexible, crystal-clear thermoplastic tubing engineered for laboratory and medical vacuum and fluid transfer applications. It offers broad chemical compatibility, a glassy-smooth inner bore to minimize buildup and carryover, and vacuum performance suitable for many analytical and diagnostic systems. Available sizes and vacuum ratings vary by specific dimension; consult the manufacturer's specifications for exact values.
  • Crystal-clear for visual inspection and bubble detection.
  • Suitable for vacuum applications up to 29.9 inHg (760 mmHg) for many sizes.
  • Thermoplastic material, non-oxidizing and non-contaminating.
  • Glassy-smooth inner bore reduces buildup and carryover.
  • Available in a wide range of inner and outer diameters; refer to specifications for exact dimensions.
  • Phthalate- and BPA-free formulation.