Learn More
Saint Gobain Performance Plastics Corp Versilon™ 3/8" ID x 1/2" OD x 1/16" Wall 100' Package Length (C-219-A) - Flexible PVC Tubing
Supplier:  Saint Gobain Performance Plastics Corp ABH02027
Encompass_Preferred
Formula C-219-A Tubing. Clear, flexible tubing74°C (165°F) maximum operating temperatureTensile strength of 2450psi (16.9MPa)
Catalog No. 50-206-7710
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product