Learn More
Accuflow Systems Saint-Gobain Vincon Flexible PVC Tubing, 3/8 in ID, 100 ft
Supplier: Accuflow Systems ABH02027
Versilon Vincon flexible PVC tubing is a clear, flexible fluid-transfer tubing formulated with a non-DEHP plasticizer for broad chemical compatibility and extended flex life. Supplied in 100 ft coils, it is suitable for laboratory and industrial fluid-handling applications such as peristaltic pumps, reagent transfer, and detergent handling.
- Clear flexible PVC material with non-DEHP plasticizer.
- Internal diameter 3/8 in, outer diameter 1/2 in, wall thickness 1/16 in.
- Coil length 100 ft.
- Maximum recommended operating temperature 74 °C (165 °F).
- Low temperature flexibility to -43 °C (-45 °F).
- Durometer hardness Shore A 67; tensile strength 2,450 psi (16.9 MPa).
- Suitable for peristaltic pumps, fluid transfer, and detergent handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.