Learn More
Accuflow Systems TUBING VINYL 1/2 x 1/8 50FT/PK
Catalog No. 502069019
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 1/2 in. 12.7 mm Diameter Outer 3/4 in. 19 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9019 Supplier Accuflow Systems Supplier No. AGL00038
Saint Gobain Clear PVC Tubing Diameter Inner 1/2 in. 12.7 mm Diameter Outer 3/4 in. 19 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m