Learn More
Accuflow Systems Clear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter, 50 FT/per pack
Catalog No. 502069011
Encompass_Preferred
This item has a minimum order quantity of 42 packs per supplier requirements. Saint Gobain Clear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 3/8 in. 9.5 mm Thickness 1.6 mm 0.0625 in. Max. Operating Temperature 165 deg.F 73.8 deg.C Overall Length 50 ft. 15.2 m Packaging Quantity 1/Pk of 50 feet.
This item is not returnable. View return policy
Catalog No. 50-206-9011 Supplier Accuflow Systems Supplier No. AGL00017
This item has a minimum order quantity of 42 packs per supplier requirements. Saint Gobain Clear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 3/8 in. 9.5 mm Thickness 1.6 mm 0.0625 in. Max. Operating Temperature 165 deg.F 73.8 deg.C Overall Length 50 ft. 15.2 m Packaging Quantity 1/Pk of 50 feet.