Learn More
Accuflow Systems TUBING VINYL 1/4 x 1/8 50FT/PK
Supplier:  Accuflow Systems AGL00019
Encompass_Preferred
Saint GobainClear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 1/2 in. 12.7 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9013