Learn More
Accuflow Systems TUBING VINYL 1/4 x 3/32 50FT/PK
Catalog No. 502069012
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 7/16 in. 11 mm Thickness 2.4 mm 0.093 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9012 Supplier Accuflow Systems Supplier No. AGL00018
Saint Gobain Clear PVC Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 7/16 in. 11 mm Thickness 2.4 mm 0.093 in. Overall Length 50 ft. 15.2 m