Learn More
Accuflow Systems TUBING VINYL 1/8 x 1/16 50FT/PK
Catalog No. 502069009
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 0.12 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m
This item is not returnable. View return policy
Catalog No. 50-206-9009 Supplier Accuflow Systems Supplier No. AGL00007
Saint Gobain Clear PVC Tubing Diameter Inner 0.12 in. 3.2 mm Diameter Outer 1/4 in. 6.4 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m