Learn More
Accuflow Systems TUBING VINYL 3/16 x 1/16 50FT/PK
Catalog No. 502069010
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 0.188 in. 4.8 mm Diameter Outer 5/16 in. 7.9 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9010 Supplier Accuflow Systems Supplier No. AGL00012
Saint Gobain Clear PVC Tubing Diameter Inner 0.188 in. 4.8 mm Diameter Outer 5/16 in. 7.9 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m