Learn More
Accuflow Systems TUBING VINYL 3/4 x 1/8 50FT/PK
Supplier:  Accuflow Systems AGL00053
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 3/4 in. 19 mm Diameter Outer 1 in. 25.4 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9021