Learn More
Accuflow Systems TUBING VINYL 3/8 x 1/16 50FT/PK
Catalog No. 502069015
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 3/8 in. 9.5 mm Diameter Outer 1/2 in. 12.7 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m
This item is not returnable. View return policy
Catalog No. 50-206-9015 Supplier Accuflow Systems Supplier No. AGL00027
Saint Gobain Clear PVC Tubing Diameter Inner 3/8 in. 9.5 mm Diameter Outer 1/2 in. 12.7 mm Thickness 1.6 mm 0.0625 in. Overall Length 50 ft. 15.2 m