Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, Flexible Vinyl, 3/8"" ID x 1/2"" OD; 50 ft
Catalog No. NC9780522
Encompass
The tubing is ideal for general-purpose lab use. Characteristics include fair chemical and abrasion resistance, odorless, nontoxic, and transparent. Tubing is manufactured according to GMP and has a temperature range of -45 to 180¿F (-43 to 82¿C). Can be sterilized with Ethylene oxide.
Catalog No. NC9780522 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0640512
The tubing is ideal for general-purpose lab use. Characteristics include fair chemical and abrasion resistance, odorless, nontoxic, and transparent. Tubing is manufactured according to GMP and has a temperature range of -45 to 180¿F (-43 to 82¿C). Can be sterilized with Ethylene oxide.