Learn More
Accuflow Systems TUBING VINYL 3/8 x 1/8 50FT/PK
Catalog No. 502069016
Encompass_Preferred
Saint Gobain Clear PVC Tubing Diameter Inner 3/8 in. 9.5 mm Diameter Outer 5/8 in. 15.9 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m
Catalog No. 50-206-9016 Supplier Accuflow Systems Supplier No. AGL00029
Saint Gobain Clear PVC Tubing Diameter Inner 3/8 in. 9.5 mm Diameter Outer 5/8 in. 15.9 mm Thickness 3.2 mm 0.125 in. Overall Length 50 ft. 15.2 m