Learn More
Perkin Elmer US LLC Three-Stop PVC Peristaltic Pump Tubing, 2.06 mm I.D., Purple/Purple/Purple, Pkg. 12
Catalog No. 509052000
Encompass_Preferred
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. This clear tubing ships as a package of 12.
Catalog No. 50-905-2000 Supplier Perkin Elmer US LLC Supplier No. B0199034
This Three-Stop Peristaltic Pump Tubing is designed for use with the FIAS, FIMS, and the S10 Autosampler. This clear tubing ships as a package of 12.