Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, FDA-Compliant Viton, L/S 18; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9641218/DEL
Encompass
Applications: Food or laboratory applications where FDA compliance is required, acid and solvent transfer, high-temperature work Characteristics: Good chemical resistance, opaque black.
Catalog No. NC9957081