Learn More
Hach Company Tubing With Anti-Diffusion Tip for AT Titrator - Delivery Tubing Accessory
Catalog No. NC3368682 Shop All Hach Company Products
Encompass
Replacement delivery tubing with an anti-diffusion tip designed for use with AT series titrators. The anti-diffusion tip reduces reagent diffusion during dispensing to help maintain titration accuracy and repeatability.
  • Prevents diffusion during reagent delivery.
  • Designed for use with AT titrator series.
  • Serves as a replacement delivery tubing accessory for titration systems.
  • Helps maintain titration accuracy and repeatability.
Catalog No. NC3368682 Supplier Hach Company Supplier No. LZE112
Replacement delivery tubing with an anti-diffusion tip designed for use with AT series titrators. The anti-diffusion tip reduces reagent diffusion during dispensing to help maintain titration accuracy and repeatability.
  • Prevents diffusion during reagent delivery.
  • Designed for use with AT titrator series.
  • Serves as a replacement delivery tubing accessory for titration systems.
  • Helps maintain titration accuracy and repeatability.