Learn More
Waters Corp Tubing, Yellow, 1/16 in ID x 25 ft
Supplier:  Waters Corp WAT077040
Encompass_Preferred
The Yellow Teflon Tubing is a needle wash outlet line that goes to waste.
Catalog No. 50-793-713