Learn More
Cytoskeleton, Inc. Tubulin polymerization assay using >99% pure tubulin, fluorescence based, 96 Assays
Catalog No. NC9050409
Encompass
Tubulin polymerization assay using > 99% pure tubulin, fluorescence based (BK011P) Uses Include: Cost effective high throughput screen for anti-cancer compounds.Basic research to measure compound IC50s and specificity for tubulin. Screening proteins for effects on tubulin polymerization activity. Teaching aid for undergraduate/graduate class in pharmacology.
Catalog No. NC9050409 Supplier Cytoskeleton, Inc. Supplier No. BK011P
Tubulin polymerization assay using > 99% pure tubulin, fluorescence based (BK011P) Uses Include: Cost effective high throughput screen for anti-cancer compounds.Basic research to measure compound IC50s and specificity for tubulin. Screening proteins for effects on tubulin polymerization activity. Teaching aid for undergraduate/graduate class in pharmacology.